missing translation for 'onlineSavingsMsg'
Learn More

Manutan Polypropylene tapes and dispenser kit, brown

Product Code. 12880469
Encompass_Preferred
Click to view available options
Color:
Brown
This item is not returnable. View return policy

Product Code. 12880469

Brand: Manutan A329065

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Pistol grip dispensers with built-in brake. 25 micron brown tape. Acrylic adhesive.
  • Roll width : 48mm
  • Roll length : 66m
  • Tape colour : Brown
  • Number of rolls : 72
  • Number of dispensers : 2

Specifications

Color Brown
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.