Learn More
Gosselin™ Polypropylene Straight Specimen Containers
Product Code. 15408814
Click to view available options
Capacity (Metric):
40 mL
60 mL
missing translation for 'unitSize'
Pack of 1200
Pack of 800
Product Code. 15408814
missing translation for 'mfr': Gosselin™ TP30C030
CORNING GOSSELIN™ Straight Container, Specimen Vials, PP come in different sizes and quantities, and are available sterile and non-sterile.
  • Clear and break-resistant polypropylene (PP)
  • Available in different sizes and quantities
  • Color-coded screw caps
TRUSTED_SUSTAINABILITY
Specifications
Product Type Specimen Container
Closure Included No
Material Polypropylene
Shape Straight
Sterility Non-sterile
Capacity (Metric) 40 mL
Color Clear
Label No Label
No. per Case 1200/Cs.
No. per Pack 1200/Pk.