missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ Polypropylene Stirring Rod
Description
- Spatula-shaped extension
- For effective manual stirring of small volumes
- 245mm in length
Specifications
Specifications
| Color | White |
| For Use With (Application) | Effective manual stirring of small volumes |
| Length (Metric) | 245 mm |
| Material | Polypropylene |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?