missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Dual Ended Stirring Rods

Stir and mix liquids with the flat button end or use the rounded end to swirl. Azpack™ Dual Ended Stirring Rods are constructed from chemical resistant polypropylene.

Brand:  Azpack™ 11368113

13.70 GBP valid until 2022-10-02
Use promo code "18216" to get your promotional price.



Code : 03

Additional Details : Weight : 0.06480kg

Product Code. 11368113

  • £24.90 / Pack of 6

In Stock
to see stock.
Add to basket

Description

Description

  • Polypropylene
  • One end rounded, one end with flat button
Specifications

Specifications

Polypropylene
Stirring rod
Translucent
150mm
Cylindrical with button and rounded ends
6PK
10mm
Videos
SDS
Documents
Certificates
Special Offers

Special Offers