missing translation for 'onlineSavingsMsg'
Learn More

Biodesign™ Polypropylene Sieve Holders

Product Code. 12987477
Click to view available options
Diameter (Metric) Outer:
152 mm
203 mm
254 mm
76, 100, 152, 203, 254 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12987477

Brand: Biodesign™ F6

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with CellMicroSieves™

Unlike traditional filters, CellMicroSieves™ are a precision woven matrix of pure nylon filaments that is available with pores ranging from 5 μm to 200 μm. CellMicroSieves™ do not wet and contains no dyes, detergents or extractables. Extremely low protein affinity means that cells and proteins will not bind to CellMicroSieves™. They have a wide variety of distinctive uses a specific sized porous screens in the laboratory.

AccessoriesPolypropylene sieve holders (2 piece) holders with adjustable metal tightener or filtration units, type A borosilicate glass with Stainless Steel filter support and aluminum clamp or type B complete polysulfone filtration unit with polypropylene filter support are available. Both filtration units have a 300 mL capacity are autoclavable and reusable.Use sheets and rolls with sieve holders and circles with filtration units to hold securely when processing samples.

  • A pure nylon product that is biologically inert, resistant to most acidic or basic conditions and many solvents
  • Autoclavable and can be repeatedly washed and reused
  • Rapid collection of tissue culture cells without centrifugation
  • Isolation of specific cell types from a dissociated tissue sample, allowing only a single type of cell to pass through
  • Separation of different sized cells from a mixture of cell types
  • Concentration and isolation of cells from a bioreactor
  • Prefilter to prevent clogging of final filter

CellMicroSieves™ allow the rapid gentle isolation of cells, organelles or cellular debris.

Specifications

Quantity Each
Type Sieve Holder
Material Polypropylene
For Use With (Equipment) Cellmicrosieves
Includes Adjustable Metal Tightener
Diameter (Metric) Outer 152 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.