missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polypropylene sheet, 3.18mm (0.125in) thick, Thermo Scientific Chemicals
Click to view available options
Quantity:
300 x 300 mm
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS | 9003-07-0 |
Molecular Formula | (C3H6)n |
MDL Number | MFCD00084447 |
Synonym | oxiranedodecanoic acid, 3-octyl-, cis, 12-2s,3r-3-octyloxiran-2-yl dodecanoic acid, polypropylene, 2-oxiranedodecanoic acid, 3-octyl-, 2r,3s-rel, oxiranedodecanoic acid, 3-octyl-, 2r,3s-rel, 3beta-octyloxirane-2beta-dodecanoic acid, 12-3-octyl-2-oxiranyl dodecanoic acid #, special for nanometer polypropylene pipe material |
Quantity | 300 x 300 mm |
InChI Key | NSYDMBURIUSUDH-UHFFFAOYNA-N |
SMILES | CC(-*)C-* |
IUPAC Name | 12-[(2S,3R)-3-octyloxiran-2-yl]dodecanoic acid |
Molecular Weight (g/mol) | 354.58 |
PubChem CID | 76958 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'