missing translation for 'onlineSavingsMsg'
Learn More

JG Finneran Associates™ Polypropylene Screw Thread Closures

Product Code. 10350331
Change view
Click to view available options
Closure Size:
10-425
8-425
Unit Size:
Pack of 1000
Product Code. Closure Size unitSize
10350331 10-425 Pack of 1000
10197360 8-425 Pack of 1000
2 options
This item is not returnable. View return policy

Product Code. 10350331

Brand: JG Finneran Associates™ 531010

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Black
Material Polypropylene
Includes 10mil molded septa
Quantity 1000/Pk.
Closure Size 10-425
For Use With (Equipment) 12 x 32 Screw Thread Vials with 10 to 425 mm Neck Finish
Product Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.