Learn More
JG Finneran Associates™ Polypropylene Screw Caps
Brand: JG Finneran Associates™ 5320-10
Code : 17
Additional Details : Weight : 0.01000kg
Specifications
Linerless | |
Polypropylene | |
1000 Pk. | |
10-425 |
Black | |
Molded | |
Unlined Solid Top Screw Cap | |
Autosamplers |