missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Polypropylene Screw Caps

Product Code. 15281249 Shop All Brand Products
Click to view available options
Product Type:
Screw Cap
Tamper-evident Screw Cap
Thread Size (Metric):
GL 25
GL 32
GL 45
GL 54
GL 80
missing translation for 'unitSize'
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 15281249

missing translation for 'mfr': BRAND™ 122620

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from polypropylene. BRAND™ Polypropylene Screw Caps are available in a variety of sizes. Pouring rings sold separately.

  • Manufactured from polypropylene.
  • Replacement and spare screw caps for laboratory bottles available in a variety of sizes.
  • Pouring rings sold separately.
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material Polypropylene
Quantity 10/Pk.
Thread Size (Metric) GL 25
Certified Yes
Neck Finish 22 mm
Product Type Screw Cap
For Use With (Equipment) For use with laboratory bottles
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.