missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Polypropylene Screw Caps
Shop All Brand Products
Click to view available options
Product Type:
Screw Cap
Tamper-evident Screw Cap
Thread Size (Metric):
GL 25
GL 32
GL 45
GL 54
GL 80
Unit Size:
Each
Pack of 10
Description
- Manufactured from polypropylene.
- Replacement and spare screw caps for laboratory bottles available in a variety of sizes.
- Pouring rings sold separately.
Specifications
Specifications
| Color | Blue |
| Material | Polypropylene |
| Quantity | 100/Pk. |
| Thread Size (Metric) | GL 80 |
| Certified | Yes |
| Neck Finish | 30 mm |
| Mouth | Wide |
| Product Type | Tamper-evident Screw Cap |
| For Use With (Equipment) | For use with wide-neck bottles |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction