Learn More
Menshen™ Polypropylene Screw Caps Wide Neck, Blue
Product Code. 10441731
Click to view available options
Certifications/Compliance:
DIN 45
DIN 54
Unit Size:
Each
Product Code. 10441731
Brand: Menshen™ 21663021
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use Kautex™ Polypropylene Screw Caps Wide Neck, Blue to top off wide neck bottles used in industry or the laboratory
  • Blue
  • For 500-1000ml bottle
  • PP
Specifications
Color Blue
Material Polypropylene
No. per Pack 1 Each
Quantity 1 Each
Screw Color Blue
Screw Material Polypropylene
Closure Color Blue
Certifications/Compliance DIN 45
Product Type Screw Cap
Closure Material Polypropylene
Closure Type Screw Cap