missing translation for 'onlineSavingsMsg'
Learn More

Samco Polypropylene Screw Caps

Product Code. p-8009999
Click to view available options
For Use With (Equipment):
T101/V3 and T101/V5
T101/V5
Unit Size:
Pack of 264
Pack of 399
This item is not returnable. View return policy

Product Code. 12967836

Brand: Samco T001/C3

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Polypropylene Screw Caps, For Use With: For T101/V5, Black, Closure Type: Screw Cap, No. per Case: 264Pack, No. per Pack: 264Pack, Packaging: 264Pack, Quantity: 264Pack

Specifications

Product Type Screw Cap
Material Polypropylene
Color Black
No. per Pack 933 Pack
No. per Case 933 Pack
Packaging 933 Pack
Quantity 933 Pack
For Use With (Equipment) T101/V3 and T101/V5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.