missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Samco Polypropylene Screw Caps
Click to view available options
For Use With (Equipment):
T101/V3 and T101/V5
T101/V5
Unit Size:
Pack of 264
Pack of 399
Description
Polypropylene Screw Caps, For Use With: For T101/V5, Black, Closure Type: Screw Cap, No. per Case: 264Pack, No. per Pack: 264Pack, Packaging: 264Pack, Quantity: 264Pack
Specifications
Specifications
| Product Type | Screw Cap |
| Material | Polypropylene |
| Color | Black |
| No. per Pack | 933 Pack |
| No. per Case | 933 Pack |
| Packaging | 933 Pack |
| Quantity | 933 Pack |
| For Use With (Equipment) | T101/V3 and T101/V5 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction