missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Screw Caps for Microtubes

Product Code. 10354101 Shop All Brand Products
Click to view available options
Color:
Blue
Red
White
Yellow
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 10354101

Brand: Brand™ 780741

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from polypropylene with silicone seal. BRAND™ Screw Caps for Microtubes are available in a variety of colors. For use at temperatures between –196 to 121°C. Autoclavable.

  • Manufactured from polypropylene with silicone seal.
  • Available in a variety of colors.
  • Temperature range: –196 to 121°C.
  • Autoclavable;
TRUSTED_SUSTAINABILITY

Specifications

Product Type Screw Cap with Silicone Seal
Autoclavable Autoclavable
Material Polypropylene
Color Blue
For Use With (Equipment) Bulk Screw Cap Microtubes 0.5 to 2 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.