Learn More
Buerkle™ 2-Part Shovel for foodstuffs
Product Code. 10521142 Shop All Bürkle Products
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Each
Product Code. Material Unit Size
10521142 Polypropylene Each
1 options
Product Code. 10521142 Supplier Buerkle™ Supplier No. 83002010
  • ONLINE EXCLUSIVE PRICE
    £61.25
    £60.15 / Pack of 1
    Save £1.10
    1% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Polypropylene's easy-to-clean, smooth and pore-free surface makes these scoops extremely suitable for the food sector. The scoops can be also used for boiling products.
  • For foodstuffs (FDA and (EU) Nr. 10/2011)
  • Easy-to-clean, smooth and pore-free surface
  • The scoops can be also used for boiling products
TRUSTED_SUSTAINABILITY
Specifications
Product Type Two-Part Shovel
Material Polypropylene
Depth (Metric) 315 mm
Includes Scraper With Fixed Handle, Hand Shovel, Filling Scoop For Ideal For Sampling, Scoop Two-Piece With D-grip, Two-Piece Shovel With T-Handle, Shovel In One Piece For Reduced Contamination
Length (Metric) 966 mm
Width (Metric) 247 mm