missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Essentials Hinged-Lid Sample Containers, PP

Product Code. 11720379 Shop All Cole-Parmer Products
Change view
Click to view available options
Capacity (Metric):
7.5 mL
Unit Size:
Pack of 2500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11720379 7.5 mL Pack of 2500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11720379 Supplier Cole-Parmer™ Essentials Supplier No. 6609800
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

The hinged cap design of these Polypropylene Sample Containers allows you to easily close the lid with one finger.

  • FDA compliant.
  • Autoclavable.
  • Made from polypropylene (PP), thess continaers are ideal for storing liquids or solids and for metals analysis.
  • When containers are properly closed, your sample won't spill or leak in any position, reducing the chance of contamination.

Specifications

Capacity (Metric) 7.5 mL
Diameter (Metric) Outer Neck 38 mm
Material Polypropylene
No. per Case 2500 Pk.
Certifications/Compliance FDA Compliant
Closure Type Hinged
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.