Learn More
BRAND™ Polypropylene Reservoir
Reservoirs allow for different volumes and different solutions to be used in the automated liquid handling process.
Brand: BRAND™ 701450
Additional Details : Weight : 0.55640kg
Description
- Pyramidal or flat bottom
- DNase-free
- RNase-free
Specifications
DIN EN 285 | |
220 mL/Column | |
Non-sterile | |
Yes | |
Conical |
96-well | |
Polypropylene | |
220 mL | |
1 |