Learn More
Ismatec™ Reducer Tubing Connectors, Plastic
Variously sized and shaped reducer tubing connectors. Ismatec™ Reducer Tubing Connectors, Plastic are available in different plastic compositions.
Brand:  Ismatec™ ISM569A
 View more versions of this product
Product Code. 15295619
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Reducer tubing connector, with varying inlet and outlet diameters.
  • Designed for Ismatec™ pump tubing.
  • Available in different sizes, shapes, and compositions.
Specifications
Specifications
Reducer Tube Connector
For 1.5mm and 2.5mm I.D. Tubing
1.5mm and 2.5mm I.D.
Polypropylene
10/Pk.
Straight