missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Polypropylene Racks

Brand:  Tenak™ TE22390

Additional Details : Weight : 0.34000kg

 View more versions of this product

Product Code. 15210584

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Specifications

Specifications

Blue
Rack
25 (15 mL tubes)
130 mm
Polypropylene
For 15 and 50mL Centrifuge Tubes
12 Pack
130 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions