Learn More
Brand™ Polypropylene Pouring Rings
Shop All Brand Products
Click to view available options
Thread Size (Metric):
GL45
Unit Size:
Pack of 10
Description
- Manufactured from polypropylene.
- For use with GL45 thread laboratory bottles.
Specifications
Specifications
| Material | Polypropylene |
| Quantity | 10Pack |
| Thread Size (Metric) | GL45 |
| Neck Finish | 22 mm |
| Product Type | Pouring Ring |
| For Use With (Equipment) | Fits GL 45 thread laboratory bottles |