missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Pouring Ring

Product Code. 11597033
Change view
Click to view available options
Color:
Yellow
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11597033 Yellow Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11597033 Supplier Fisherbrand™ Supplier No. E5004/05

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

For Use With (Application) Reagent and media bottles
Product Type Pouring Ring
Material Polypropylene
Color Yellow
Size 45 GL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.