missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Socorex™ Polypropylene Pipette Tips
Description
- Supplied in loose in bags or racks with lids
- Autoclavable at 121°C / 250°F
- Compatible with other brands' tips
Specifications
Specifications
| Volume (Metric) | 350 μL |
| Color | Natural |
| Autoclavable | Autoclavable |
| Format | Rack |
| Tip Style | Universal |
| Packaging | Racked, 960/pk |
| For Use With (Equipment) | Socorex, Eppendorf, Gilson |
| Material | Polypropylene |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?