missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Polypropylene Pipette tips

Product Code. 10451243
Change view
Click to view available options
Volume (Metric):
0.2 to 10 μL
1 to 250 μL
100 to 1200 μL
5 to 300 μL
50 to 1200 μL
Format:
Bulk
Individually Wrapped
Unit Size:
Pack of 1000
Pack of 120
Pack of 400
Pack of 960
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Format unitSize
10451243 1 to 250 μL Individually Wrapped Pack of 1000
10591153 5 to 300 μL Individually Wrapped Pack of 1000
10141191 50 to 1200 μL Individually Wrapped Pack of 400
10518581 0.2 to 10 μL Bulk Pack of 960
10213721 1 to 250 μL Bulk Pack of 960
10727471 100 to 1200 μL Bulk Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10451243 Supplier Hirschmann™ Supplier No. 9490106

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Constructed from polypropylene, for use with the labopette™

  • Free of cadium
  • Certified conformity
  • Polypropylene
  • Specifications

    Description Cadmium-free
    Volume (Metric) 1 to 250 μL
    Color Clear
    Certifications/Compliance Conformity certified (when being used with micro pipettes labopette™)
    Format Individually Wrapped
    Packaging Bulk packed in bag/carton
    Quantity 1000/Pack
    Product Type Tips
    For Use With (Equipment) Hirschmann Laborgerate (For Multichannel Pipette)
    Material Polypropylene
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.