missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Polypropylene Pipette Tips

Product Code. 12533287
Click to view available options
Volume (Metric):
0.1 to 10 mL
0.5 to 20 μL
1000 to 5000 μL
2 to 300 μL
Format:
Bulk
Rack
Refill
Color:
Light Blue
Natural
Neutral
Unit Size:
Pack of 1000
Pack of 200
Pack of 4800
Pack of 960
This item is not returnable. View return policy

Product Code. 12533287

Brand: Kartell™ 0097100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Reduce sample retention with a tip that offers high clarity and excellent water-repellent properties. Kartell™ Polypropylene Pipette Tip uses precise cerntering to guarantee reliable directionality of the sample when delivered.

  • High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
  • The precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
  • These tips are recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market
  • Disposable tips are also available racked

Specifications

Volume (Metric) 2 to 300 μL
Color Neutral
Autoclavable Autoclavable
Certifications/Compliance CE / IVD
Format Bulk
For Use With (Equipment) Kartell, Biohit, Gilson, Finnpipette, Labsystem
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.