missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Polypropylene Pipette Tips

Product Code. 10611304
Change view
Click to view available options
Volume (Metric):
200 μL
350 μL
1000 μL
5 mL
Color:
Blue
Natural
Yellow
Format:
Bulk
Rack
Unit Size:
Pack of 100
Pack of 1000
Pack of 480
Pack of 960
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color Format unitSize
10611304 5 mL Natural Rack Pack of 100
10171412 350 μL Natural Bulk Pack of 1000
10447282 350 μL Natural Rack Pack of 960
11763266 200 μL Yellow Rack Pack of 960
11783246 1000 μL Blue Rack Pack of 480
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10611304 Supplier Socorex™ Supplier No. 312.05R
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use Socorex™ Polypropylene Pipette Tips when a whole volume range since there is a whole volume range from which to choose. They are available in various colors, are constructed of polypropylene and are autoclavable for cost effectiveness.

  • Supplied in loose in bags or racks with lids
  • Autoclavable at 121°C / 250°F
  • Compatible with other brands' tips

Specifications

Volume (Metric) 5 mL
Color Natural
Autoclavable Autoclavable
Format Rack
Tip Style Universal
Packaging Racked, 100/pk
For Use With (Equipment) Socorex, Eppendorf
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.