missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Polypropylene Pipet Finntip™

Product Code. 10106140
Change view
Click to view available options
Volume (Metric):
1000 to 5000 μL
2000 to 10,000 μL
Unit Size:
Pack of 100
Pack of 75
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10106140 2000 to 10,000 μL Pack of 100
10130101 1000 to 5000 μL Pack of 75
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10106140 Supplier Ratiolab™ Supplier No. 2400640
Only null left
Add to basket
Add to basket

Use these pipet tips for simple and safe liquid handling in the most sensitive applications. Ratiolab™ Polypropylene Pipet Finntip™ are ideal for manual or electronic, single or multichannel pipets.

  • Ideal for all commercial pipets
  • Made of high quality  polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to stable shape
  • Autoclavable
  • Guaranteed dust free due to special ionization treatment
  • Excellent transparency and high quality surfaces
  • Minimal residual wetting
  • Excellent volume accuracy due to precise tip orifice
  • Practice-oriented tip volumes

Specifications

Volume (Metric) 2000 to 10,000 μL
Color Translucent
Autoclavable Autoclavable
Length (Metric) 15.02cm
Certifications/Compliance CE, IVD
For Use With (Application) Ideal for All Commercially Available Pippets
Sterilization Method Autoclavable
For Use With (Equipment) Universal Fit
Product Line FinnTip
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.