Description
- Kit contains 10 x filter 2 x Support screen
- The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, except 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran
- Polypropylene (PP)
Specifications
Specifications
| Temperature (Metric) Max | 4°C |
| Packaging Quantity | 10 Pack |