Learn More
Mutexil™ Visitor Protective Lab Coats
Product Code. 11700524
Click to view available options
Closure Type:
Snaps
Size:
X-Large
2X-Large
Unit Size:
Pack of 200
Pack of 50
Product Code. 11700524
Brand: Mutexil™ 4574739XXL
Only null left
Add to basket
Add to basket
Use Mutexil™ Visitor Protective Lab Coats for visitors' safety and protection when visiting a facility that merits this required protective clothing.
PP non woven labcoat with press-stud fastening and pockets
  • Disposable
  • PP, 40g/m2
  • For maintenance, production site visits
  • For non hazardous environments
Specifications
Product Type Lab Coat
Size 2X-Large
Color White
Closure Type Snaps
No. of Pockets None
Packaging Type Individually bagged
Construction Non-woven