missing translation for 'onlineSavingsMsg'
Learn More

Brady™ MiniMark™ Polypropylene Unprinted Labels

Product Code. 11797324
Click to view available options
Width (Metric):
55 mm
75 mm
105 mm
Height (Metric):
85.00 mm
110.00 mm
155.00 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11797324

Brand: Brady™ 710678

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MiniMark™ Polypropylene Unprinted Labels are ready for you to print the required warning compliant to the latest regulations.

  • Unprinted labels ready for you to print the required warning compliant to the latest regulations.

Specifications

Certifications/Compliance UL Recognised
Description Unprinted MiniMark Labels for CLP-GHS Hazardous Substances
Adhesive Type Acrylic, Permanent
Coating None
Color White
Display Caution
Finish Gloss
Format Label
Material Polypropylene
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Hazard Communication Identification
Labels per Row 1
Min. Operating Temperature (Metric) -20°C
No. per Pack 300 Labels/Box
Quantity 300 Pk.
Shape Rectangle
Temperature Range (English) -4°F to +212°F
Temperature Range (Metric) -20°C to +100°C
Thickness (English) 0.0031 in.
Thickness (Metric) 0.079 mm
Vertical Repeat (Metric) 115.36 mm
Dimensions (LxW) 110 x 75 mm
Length (English) 4.33 in.
Length (Metric) 110 mm
Width (English) 2.95 in.
Width (Metric) 75 mm
Height (Metric) 110.00 mm
Alternate Part Number MNK-75x110-B7594-0.3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.