missing translation for 'onlineSavingsMsg'
Learn More

Brady™ MiniMark™ Polypropylene Unprinted Labels

Product Code. 11707344 Shop All Brady Products
Change view
Click to view available options
Width (Metric):
55 mm
Height (Metric):
85.00 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Height (Metric) Unit Size
11707344 55 mm 85.00 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11707344 Supplier Brady™ Supplier No. 710676
  • ONLINE EXCLUSIVE PRICE
    £144.00
    £140.10 / Pack of 1
    Save £3.90
    2% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MiniMark™ Polypropylene Unprinted Labels are ready for you to print the required warning compliant to the latest regulations.

  • Unprinted labels ready for you to print the required warning compliant to the latest regulations.

Specifications

Certifications/Compliance UL Recognised
Description Unprinted MiniMark Labels for CLP-GHS Hazardous Substances
Adhesive Type Acrylic, Permanent
Coating None
Color White
Display Blank
Finish Gloss
Format Label
Material Polypropylene
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) High performance material designed to perform on various surfaces
Min. Operating Temperature (Metric) -20°C
No. per Pack 400 Labels/Box
Quantity 400 Pk.
Temperature Range (English) -4°F to +212°F
Temperature Range (Metric) -20°C to +100°C
Thickness (English) 0.0031 in.
Thickness (Metric) 0.079 mm
Vertical Repeat (Metric) 91.02 mm
Dimensions (L x W) 85 x 55 mm
Length (English) 3.35 in.
Length (Metric) 85 mm
Width (English) 2.17 in.
Width (Metric) 55 mm
Height (Metric) 85.00 mm
Alternate Part Number MNK-55x85-B7594-0.4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.