missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Microcentrifuge Tubes Without Screw Caps

Product Code. 10304281 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
2 mL
Format:
Conical bottom
Self-standing, conical bottom
Unit Size:
Pack of 1000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Format unitSize
10304281 1.5 mL Self-standing, conical bottom Pack of 1000
10529701 0.5 mL Self-standing, conical bottom Pack of 1000
10461013 1.5 mL Conical bottom Pack of 1000
10354371 2 mL Conical bottom Pack of 1000
10539891 2 mL Self-standing, conical bottom Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10304281 Supplier BRAND™ Supplier No. 780731
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from polypropylene and complies with DIN EN 285 standards. BRAND™ Microcentrifuge Tubes Without Screw Caps are graduated with frosted marking area that allows for sample identification or other notations. Colored screw caps and cap inserts available for separate purchase.

  • Manufactured from polypropylene and complies with DIN EN 285 standards.
  • Graduated.
  • Frosted marking area allows for sample identification or other notations.
  • Operating range: -196°C to 121°C.
  • Autoclavable at 121°C (2 bar)
  • Colored screw caps and cap inserts available for separate purchase.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance DIN EN 285
Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Material Polypropylene
Format Self-standing, conical bottom
No. per Pack 1000/Pk.
Temperature Range (Metric) –196 to 121°C
Sterility Non-sterile
Graduated Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.