missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Polypropylene Micro Pipet Tips

Product Code. 10003261
Change view
Click to view available options
Sterility:
Non-sterile
Quantity:
5000/Pack
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Quantity unitSize
10003261 Non-sterile 5000/Pack Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10003261 Supplier Greiner Bio-One Supplier No. 771290

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed for precise pipetting of the smallest possible liquid volumes. Greiner Bio-One™ Polypropylene Micro Pipet Tips provide a small dead volume due to their small size.

  • Manufactured from high-grade polypropylene for optimum performance and fit
  • High breaking strength, dimensionally stable
  • Heat resistant to +140°C 
  • Free of heavy metals
  • Available in bulk pacaging or convenient rack
  • Conform to ISO 9001 standard
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.5 to 10 μL
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Model P10 standard
Quantity 5000/Pack
For Use With (Equipment) Gilson Pipetman U10, Gilson Pipetman P10, Gilson Pipetman P2
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.