Learn More
Masterflex™ Polypropylene Luer Fitting
Product Code. 15305641
Change view
Click to view available options
For Use With:
Female Luer x 1/4 to 28 in. UNF
Female Luer x Female Luer, Elbow
Female x Female x Female
Male LL to 1/4 to 28 UNF Thread
Male LL to 10 to 32 Thread
Male x Female x Female
Unit Size:
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
15305641 Female Luer x Female Luer, Elbow Pack of 25
15114865 Female Luer x 1/4 to 28 in. UNF Pack of 25
15116116 Female x Female x Female Pack of 25
15102954 Male LL to 1/4 to 28 UNF Thread Pack of 25
15395631 Male LL to 10 to 32 Thread Pack of 25
15329612 Male x Female x Female Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15305641 Supplier Masterflex™ Supplier No. 4550826
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Featuring a variety of fitting options, these Luer fittings are ideal for many applications including syringes or small tubing. Cole-Parmer™ Polypropylene Luer Fitting are available with standard, wide, and low-profile barbs.
  • Three barb styles: standard, wide-bore, and low-profile.
    • Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing.
    • Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene.
  • Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable.
  • All parts meet tight dimensional tolerances for increased reliability and process purity.
  • Choose from a range of sizes, 1/16in. to 1/4in., with a variety of connection options.
  • Pack of 25
Specifications
Certifications/Compliance ISO 594-1 standards
Material Polypropylene
For Use With Female Luer x Female Luer, Elbow
Packaging Quantity 25 Pack
Product Type Luer Fitting