missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Polypropylene Label Tape for BradyJet J4000

Product Code. 17459222
Click to view available options
Width (Metric):
57.1 mm
101.6 mm
152.4 mm
203.2 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17459222

Brand: Brady™ 152784

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

This B-7425J polypropylene material is ideal for GHS and other full-color identification applications where resistance to harsh chemicals is required.

  • Matte white polypropylene with acrylic adhesive is Inkjet printable
  • Low-cost, full-color GHS labeling option withstands chemical exposure
  • Service temperature of -80°C to 70°C
  • An ink cartridge is required to print this material

Specifications

Autoclavable Autoclavable
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Continuous
Material Polypropylene (Inkjet-receptive)
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Hazard Communication Identification, Equipment Labeling, Laboratory Labeling, General Identification
For Use With (Equipment) BradyJet J4000 Color Label Printer
Labels per Row 1
Min. Operating Temperature (Metric) -80°C
No. per Pack 1 Tape/Bag
Quantity 1 Tape
Temperature Range (English) -112°F to +158°F
Temperature Range (Metric) -80°C to +70°C
Thickness (English) 0.005 in.
Thickness (Metric) 0.13 mm
Dimensions (LxW) 30.48 m x 101.6 mm
Length (English) 100 ft.
Length (Metric) 30.5 m
Width (English) 4 in.
Width (Metric) 101.6 mm
Alternate Part Number J40C-4000-7425J
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.