Learn More
Chemsplash Polypropylene Lab/Visitors Coat White with Velcro Closure

Description
Visitors Polypropylene Disposable Lab Coat
- Material: 30gsm Polypropylene Fabric
- Features:
- Velcro Closure
- With Fold Over Collar
- Elasticated Cuffs
- Ideal for Non-Hazardous Environments
- Ideal for Food Industry
- PFAS Free
The Visitors Polypropylene Disposable Lab Coat is designed with a velcro front closure and is suitable for visitors, site inspections, or general wear in minimal risk environments. It is ideal for use in the food industry or other non-hazardous settings. This product is suitable for use in BRC-approved work areas and the pharmaceutical industry.
This product does not contain: PFAS, RoHS, POP, TSCA, REACH, or any conflict materials.
Specifications
Specifications
| Product Type | Visitor Lab Coat |
| Size | Small |
| Gender | Unisex |
| Material | Polypropylene |
| Color | White |
| Closure Type | Hook and Loop |
| Cuff Style | Elastic |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Resistance | Particulates |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.