missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Polypropylene: ISO Safety Sign - Wear head protection

Sign compliant to ISO 7010.

Brand:  Brady™ 819917

Additional Details : Weight : 0.19200kg

 View more versions of this product

Product Code. 15684517

  • £38.65 / Each
Estimated Shipment: 13-03-2024
to see stock.
Add to basket

Description

Description

  • Polypropylene
  • For indoor and outdoor use
Specifications

Specifications

ISO 7010
1 Ea.
Indoor and Outdoor
-40°C to 120°C
Global
Polypropylene
400 mm
1.501 mm
Round
White on Blue
Recommendations

Recommendations

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers