missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Polypropylene: ISO Safety Sign - Direction arrow 45°, Top Right Corner, Red

Sign compliant to ISO 7010.

Brand:  Brady™ 830832

Additional Details : Weight : 0.09200kg

 View more versions of this product

Product Code. 15644797

  • £14.20 / Each
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

  • Polypropylene
  • For indoor and outdoor use
Specifications

Specifications

ISO 7010
Indoor and Outdoor
1.501 mm
-40°C to 120°C
Global
Polypropylene
250 mm
250 mm
Square
White on Red
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers