missing translation for 'onlineSavingsMsg'
Learn More

Polypropylene glycol 400

Product Code. p-7051225
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11315097

Brand: Thermo Scientific Alfa Aesar 040811.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25322-69-4 | (C3H6O)n | 134.18 g/mol

It is used in cosmetic and pharmaceutical preparations and in the manufacture of emulsifying or wetting agents and lubricants,suppress foaming in industrial processes and for making polyurethane resins, hydraulic fluids, and various other materials. It acts as swelling agent for the preparation and characterization of organo-modified SBA-15. Polypropylene glycol and tetraethoxysilane precursors play an important role for the preparation of superhydrophobic silica-based surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-69-4
Molecular Formula (C3H6O)n
Molecular Weight (g/mol) 134.18
MDL Number MFCD00084446
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
Synonym polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
PubChem CID 32881
IUPAC Name 2-(2-hydroxypropoxy)propan-1-ol
SMILES [H]OC(C)CO

Specifications

Physical Form Liquid
Solubility Information Insoluble in cold water.
Quantity 50 g
Chemical Name or Material Polypropylene glycol 400

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.