missing translation for 'onlineSavingsMsg'
Learn More

Polypropylene glycol 3,000

Product Code. 11355097
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11355097 50 g 50g
11365097 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11355097 Supplier Thermo Scientific Alfa Aesar Supplier No. 040814.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25322-69-4 | (C3H6O)n | 134.18 g/mol

Propylene glycol is used in the plastics industry for the manufacture of polyester fibers and alkyd resins. It is used as a main ingredient in automobile antifreeze and engine-cooling liquids and in brake and hydraulic fluids. It is used as a humectant in foods and cosmetics. It is used as a solvent for coloring or flavoring agents as well as in many oral, injectable or topical pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-69-4
Molecular Formula (C3H6O)n
Molecular Weight (g/mol) 134.18
MDL Number MFCD00084446
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
Synonym polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
PubChem CID 32881
IUPAC Name 2-(2-hydroxypropoxy)propan-1-ol
SMILES [H]OC(C)CO

Specifications

Physical Form Liquid
Solubility Information Soluble in ethyl alcohol,ethyl ether.
Quantity 50 g
Chemical Name or Material Polypropylene glycol 3,000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.