Learn More
Hirschmann™ 96-well Microtitration Plates, PP
Click to view available options
Total Volume (Metric) Well:
1.2 mL
Unit Size:
Each
Description
- Available with fixed or loosely inserted glass inserts
- Highly pure
- Chemically inert
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Material | Glass, Polypropylene |
| No. of Wells | 96 |
| Total Volume (Metric) Well | 1.2 mL |
| Includes | Transparent cover and 96 Cavities |
| No. per Pack | 1 Each |
| Quantity | Each |