missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Depth Filter

Protects and prolongs the life of RO membranes. Fisher Scientific™ Polypropylene Depth Filters are for use with Accu500 Ultrapure Water Systems.

Brand:  Fisherbrand™

Additional Details : Weight : 2.60000kg

 View more versions of this product

Product Code. 15419649

  • £64.65 / Pack of 12
Estimated Shipment: 06-09-2024
to see stock.
Add to basket

Description

Description

  • Available in two pore sizes: 1μm and 10μm
  • Protects RO membranes
  • Filters microorganisms and particles from tap water
Specifications

Specifications

Pore size: 10μm
Accu500 Ultrapure Water System
254 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers