Learn More
Buerkle™ Polypropylene Hose Connector Box
Product Code. 10065150 Shop All Bürkle Products
Change view
Click to view available options
Packaging Quantity:
5/Scale, 68/Set
Unit Size:
Each
Product Code. Packaging Quantity Unit Size
10065150 5/Scale, 68/Set Each
1 options
Product Code. 10065150 Supplier Buerkle™ Supplier No. 87151000
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Find the appropriate hose connector you seek in this Buerkle™ Polypropylene Hose Connector Box. Included are PP Straight, Universal, T-type, Y-type, L-type, reduction adapters and two each of every size.of hose connector.
  • 68 Hose connectors
  • Polypropylene
  • Shatterproof
  • Transparent
Two of each in sizes 4, 6, 8, 10, 12 and 14 mm
  • Straight hose connector
  • T hose connector
  • Y hose connector
  • X hose connector
  • Angle hose connector
Each for hoses from 4-17 mm
  • Universal hose connector
Two of each in sizes 4-8, 4-12 and 8-12 mm
  • Reducing hose connector
  • PP, shatterproof and transparent
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EU Foodstuffs
Material Polypropylene
Packaging Quantity 5/Scale, 68/Set
Includes 56 hose connectors
Color Transparent
Product Type Hose Connector Set