missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Polypropylene Clear Disposal Bags

Product Code. 10023281
Change view
Click to view available options
Length (Metric):
500 mm
780 mm
1100 mm
Width (Metric):
300 mm
600 mm
700 mm
Capacity (Metric):
10 L
65 L
130 L
Quantity:
350/Pk.
500/Pk.
Unit Size:
Pack of 350
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Capacity (Metric) Quantity Unit Size
10023281 780 mm 600 mm 65 L 500/Pk. Pack of 500
10420722 1100 mm 700 mm 130 L 350/Pk. Pack of 350
10124581 500 mm 300 mm 10 L 500/Pk. Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10023281 Supplier Greiner Bio-One Supplier No. 646201
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Made of polypropylene foil for sterilization in the steam autoclave. Greiner Bio-One™ is ideal for the hygienic disposal of contaminated laboratory items.

  • Oxygen transport rate of 50 μm PP foil (+23°C): 2380 cm³/(m² x 24h x bar)
  • Water vapour transport rate of 50 μm PP foil (+23°C, 85% air humidity): 1.54 g/(m² x 24h)
  • Available on request with imprint Biohazard if a sufficient number is ordered
  • Foil thickness: 0.05mm
  • Autoclavable
    • Steam autoclaves: Max 134°C, 2bar/30min
    • Hot-air sterilizers: Max 140°C, 120min
  • Temperature range: 0°C to 140°C
  • For single use only
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Closure Type None
Color Translucent
Disposable Single-use
For Use With (Application) Ideal for the hygienic disposal of contaminated laboratory items
Material Polypropylene
Sterility Non-sterile
Quantity 500/Pk.
Length (English) 30.7 in.
Width (English) 23.6 in.
Product Type Autoclave Bags
Capacity (Metric) 65 L
Length (Metric) 780 mm
Width (Metric) 600 mm
Thickness (Metric) 0.050 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.