missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Features
- Base material coarse-grained kieselguhr (also known as diatomaceous earth, hydromatrix, celite)
- large pore size, high pore volume, constantly high batch-to-batch quality
- pH working range 1–13
Application
- Liquid-liquid extraction of highly viscous aqueous solutions such as physiological fluids (blood, plasma, and serum) in clinical chemistry, dyes in textiles, environmental and food analysis without use of a separation funnel
- High water loadability without breakthrough of water during elution with organic solvents
- Also suited for removing small amounts of water from solvents which are not miscible with water
Advantages
- Fast, reproducible and economical
- Simultaneous preparation of several samples
- No problems with phase separation no formation of emulsions
- High recovery rates
- Saving of time and solvents
- Organic solutions need not to be dried after separation
Solvents applicable for elution
- diethyl ether
- tert-butyl methyl ether
- ethyl acetate
- n-hexane
- cyclohexane
- toluene
- dichloromethane (methylene chloride)
- trichloromethane (chloroform)
- solvent mixtures:
- trichloromethane – methanol (90:10, v/v)
- trichloromethane – methanol (85:15, v/v)
- diethyl ether – ethanol (90:10, v/v)
- diethyl ether – ethanol (80:20, v/v)
- dichloromethane – 2-propanol (90:10, v/v)
- dichloromethane – 2-propanol (85:15, v/v)
Specifications
Specifications
| Quantity | 1000 g |
| Product Type | XTR Adsorbent |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?