missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Essentials Polypropylene Cap for E-Vac Aspiration System Vacuum Bottles
Click to view available options
Product Type:
Barbed Fitting Cap
Unit Size:
Each
Description
- Accessories for E-Vac system designed to collect critical or dangerous liquids
E-Vac aspiration systems available as 04397-10 and 13050-31, sold separately.
M.Vac Jr., polypropylene, Lid, cap
Specifications
Specifications
| Color | Natural |
| Description | M-Vac Jr. Lid with barbed tube fittings for 2L bottle |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Thread Size (Metric) | 53B |
| Disposable | Reusable |
| Product Type | Barbed Fitting Cap |
| For Use With (Equipment) | E-Vac Aspiration System Vaccum Bottles |
| Autoclavable | Not Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction