missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Polypropylene Burette Clamps

Product Code. 10375371 Shop All Brand Products
Click to view available options
For Use With (Equipment):
For use with 2 Burettes
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10375371

Brand: BRAND™ 16510

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

To be mounted on support rods of 8 to 14mm dia. Easily affixes the burette by clamping with a non-corroding spring.

  • Polypropylene construction
  • White
  • For use with support rods 8 to 14mm dia.
TRUSTED_SUSTAINABILITY

Specifications

Color White
For Use With (Equipment) For use with 2 Burettes
Material Polypropylene
No. per Pack 1 Each
Type Buret Holder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.