Learn More
Integra Biosciences™ Bottle for VACUSAFE™ Aspiration System
Click to view available options
For Use With (Equipment):
VACUSAFE Aspiration System
Unit Size:
Each
Description
- Polypropylene bottle
- 4 liters
- For use with VACUSAFE Aspiration Systems
Specifications
Specifications
| Quantity | 1 Each |
| Type | Bottle, with closed lid |
| For Use With (Equipment) | VACUSAFE Aspiration System |
| Material | Polypropylene |
| Capacity (Metric) | 4 L |