Learn More
Ratiolab™ Polypropylene Blue Pipet Tip E
Use these pipet tips for simple and safe liquid handling in the most sensitive applications. Ratiolab™ Polypropylene Blue Pipet Tips E are ideal for manual or electronic, single or multichannel pipets.
Brand: Ratiolab™ 2100610
Code : A0
Additional Details : Weight : 0.69300kg
Description
- Ideal for all commercial pipets
- Made of high quality polypropylene
- Tight fit due to the precisely fitting cone
- Optimal handling due to stable shape
- Autoclavable
- Guaranteed dust free due to special ionization treatment
- Available in pre-sterilized or non-sterile
- Excellentr4 transparency and high quality surfaces
- Minimal residual wetting
- Excellent volume accuracy due to precise tip orifice
- Practice-oriented tip volumes
Specifications
100 to 1000 μL | |
Autoclavable | |
CE, IVD | |
Autoclavable | |
Universal Fit |
Blue | |
6.9cm | |
Ideal for All Commercially Available Pippets | |
Bulk | |
Polypropylene |