missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Base Plate for Test Tube Racks

Product Code. 15835507
Click to view available options
Color:
White
Unit Size:
Pack of 4
This item is not returnable. View return policy

Product Code. 15835507

Brand: Kartell™ 0013400

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Kartell™ Base Plate for Test Tube Racks holds two test tube racks in its slots and is useful for colorimetric analysis and to stock standard samples.

  • Base plate that can hold two of Kartell™ 130 to 135 series test tube racks in the slots provided
  • Useful for colorimetric analysis and to stock standard samples

Specifications

Product Type Base Plate
Color White
Holds Test Tube Racks (Art 130 to 135 Series)
Material Polypropylene
Positions 2
For Use With (Application) Useful for colorimetric analysis and to stock standard samples
For Use With (Equipment) Kartell™ 130 - 131 - 135
Quantity 1 Pack
Dimensions (L x W x H) 156 x 202 x 13.5mm
Height (Metric) 13.5 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.