Learn More
Masterflex™ Polypropylene Barbed Connector/reducer Kits
Click to view available options
Description:
Size: Large
Size: Small
Unit Size:
Each
Description
Cole-Parmer Polypropylene Barbed Connector/reducer Kits, Autoclavable: Autoclavable, For Use With: Male Luer Lock, Packaging Quantity: 1 Pack, Barbed Connector/Reducer Kit
Specifications
Specifications
| Description | Size: Large |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| For Use With | Male Luer Lock |
| Packaging Quantity | 1 Pack |
| Product Type | Barbed Connector/Reducer Kit |